3,4,5-Trichloro-α,α,α-trifluorotoluene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,4,5-Trichlorobenzotrifluoride, 1,2,3-Trichloro-5-(trifluoromethyl)benzene
T2281-25G
T2281-5G
TCIAT2281-25GEA
575
SEK
TCIAT2281-25G
TCIAT2281-5G
3,4,5-Trichloro-α,α,α-trifluorotoluene ≥98.0%
3,4,5-Trichloro-α,α,α-trifluorotoluene
Formel:
C₇H₂Cl₃F₃ MW: 249,45 g/mol Kokpunkt: 113°C Smältpunkt: –6°C Densitet: 1,6003 g/cm³ (20 °C) Flampunkt: 98°C |
MDL Number:
MFCD00035985 CAS nummer: 50594-82-6 EINECS: 256-636-6 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...