1,14-Tetradecanedioic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Tetradecanedioic acid, 1,12-Dodecanedicarboxylic acid

T0704-25G T0704-5G
TCIAT0704-25GEA 1025 SEK
TCIAT0704-25G TCIAT0704-5G
1,14-Tetradecanedioic acid ≥98.0%
1,14-Tetradecanedioic acid
Formel: C₁₄H₂₆O₄
MW: 258,36 g/mol
Kokpunkt: 407 °C (1013 hPa)
Smältpunkt: 126 °C
Densitet: 1,108 g/cm³ (21 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002743
CAS nummer: 821-38-5
EINECS: 212-476-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5746f68ff26208921f3b11fe5db0d3a429e3e97d&vid=5746f68ff26208921f3b11fe5db0d3a429e3e97d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 125.0 to 128.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR