Dimethyl sebacate ≥98.0%
Leverantör: TCI
S0027-25G S0027-500G
TCIAS0027-25GEA 215 SEK
TCIAS0027-25G TCIAS0027-500G
Dimethyl sebacate ≥98.0%
Dimethyl sebacate
Formel: C₁₂H₂₂O₄
Kokpunkt: 102 °C (0,05 -rr)
Smältpunkt: 38 °C
Densitet: 1,0989 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008472
CAS nummer: 106-79-6
EINECS: 203-431-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c827258cdb842ba71b0bfde1d801517a3b52f1be&vid=c827258cdb842ba71b0bfde1d801517a3b52f1be&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %