(S)-(-)-2-Bromopropionic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: (S)-(-)-2-Bromopropanoic acid
B1757-25G B1757-5G
TCIAB1757-25GEA 3275 SEK
TCIAB1757-25G TCIAB1757-5G
(S)-(-)-2-Bromopropionic acid ≥98.0% (by GC, titration analysis)
(S)-(-)-2-Bromopropionic acid
Formel: H₃CBrCHCOOH
Kokpunkt: 94°C
Densitet: 1,7 g/cm³ (20 °C)
Flampunkt: 101°C
Storage Temperature: Kylskåp
MDL Number: MFCD00137325
CAS nummer: 32644-15-8
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d3125a26c028a3d108905df8d57b9cc7f1a23d3&vid=0d3125a26c028a3d108905df8d57b9cc7f1a23d3&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 95.0 ee%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -25.0 to -29.0 deg (neat)