(S)-(+)-2-Pentanol ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (S)-(+)-Propyl methyl carbinol, (S)-(+)-Methyl propyl carbinol, (S)-(+)-2-Hydroxy pentane
P0743-1ML P0743-5ML
TCIAP0743-1MLEA 1425 SEK
TCIAP0743-1ML TCIAP0743-5ML
(S)-(+)-2-Pentanol ≥98.0%
(S)-(+)-2-Pentanol
Formel: C₅H₁₂O
MW: 88,15 g/mol
Kokpunkt: 119°C
Smältpunkt: –50 °C
Densitet: 0,8169 g/cm³ (12.3 °C)
Flampunkt: 34°C
Storage Temperature: Omgivande
MDL Number: MFCD00065952
CAS nummer: 26184-62-3
UN: 1105
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c90b6faf4fa2e709c6b9b78da772d06df7d30da&vid=9c90b6faf4fa2e709c6b9b78da772d06df7d30da&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +12.0 to +15.0 deg (neat)