L(+)-Lactic acid ≥85.0%
Leverantör: TCI EUROPE
Fara
Synonyms: (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
L0165-25G L0165-500G
TCIAL0165-25GEA 245 SEK
TCIAL0165-25G TCIAL0165-500G
L(+)-Lactic acid ≥85.0%
L(+)-Lactic acid
Formel: C₃H₆O₃
MW: 90,08 g/mol
Kokpunkt: 235 °C (1013 hPa)
Smältpunkt: ∼53 °C
Densitet: ~ 1,385 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00064266
CAS nummer: 79-33-4
EINECS: 201-196-2
UN: 3265
ADR: 8,III
Merck Index: 14,05337
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7846362e1c8f4db09503ffdf99de07676ca1b00b&vid=7846362e1c8f4db09503ffdf99de07676ca1b00b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization back titration) min. 85.0 %
Specific rotation [a]20/D -10.0 to -15.0 deg (C=2.5, 1.5mol/L NaOH sol.)