(R)-(+)-2-Klorpropionsyra ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: (R)-(+)-2-Chloropropanoic acid

C1655-1G C1655-25G C1655-5G
TCIAC1655-1GEA 215 SEK
TCIAC1655-1G TCIAC1655-25G TCIAC1655-5G
(R)-(+)-2-Klorpropionsyra ≥98.0% (by GC, titration analysis)
(R)-(+)-2-Klorpropionsyra
Formel: H₃CCHClCOOH
MW: 108,52 g/mol
Kokpunkt: 185 °C (1013 hPa)
Smältpunkt: –12 °C
Densitet: 1,25 g/cm³ (20 °C)
Flampunkt: 101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064204
CAS nummer: 7474-05-7
UN: 2511
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6abb16a21d0a5e51a263537f5c8d170511a09d8c&vid=6abb16a21d0a5e51a263537f5c8d170511a09d8c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 96.0 ee%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +12.0 to +16.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR