(4-Biphenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 4-Biphenylacetic acid
B1278-25G B1278-250G
TCIAB1278-25GEA 455 SEK
TCIAB1278-25G TCIAB1278-250G
(4-Biphenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
(4-Biphenyl)acetic acid
Formel: (C₆H₅C₆H₅)CH₂COOH
MW: 212,25 g/mol
Kokpunkt: 365 °C (1013 hPa)
Smältpunkt: 160…163 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004351
CAS nummer: 5728-52-9
EINECS: 227-233-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,03316
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70a1a7a6e3648963ae602ef107ae31299da714c0&vid=70a1a7a6e3648963ae602ef107ae31299da714c0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 161.0 to 165.0 deg-C