4-Phenyl-1-butanol ≥95.0%
Leverantör: TCI EUROPE
Synonyms: 4-Phenylbutan-1-ol
P1275-25ML P1275-5ML
TCIAP1275-25MLEA 1400 SEK
TCIAP1275-25ML TCIAP1275-5ML
4-Phenyl-1-butanol ≥95.0%
4-Phenyl-1-butanol
Formel: (C₆H₅)(CH₂)₃CH₂OH
MW: 150,22 g/mol
Kokpunkt: 140°C
Densitet: 0,975 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002971
CAS nummer: 3360-41-6
EINECS: 222-128-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c55f7055592d52a187544d8dc116b723d1f265e7&vid=c55f7055592d52a187544d8dc116b723d1f265e7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %