1,2-Di(4-pyridyl)ethylene ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: (E)-1,2-Di(pyridin-4-yl)ethene, 1,2-Bis(4-pyridyl)ethylene, 4,4'-Vinylenedipyridine
D0276-10G D0276-25G
TCIAD0276-10GEA 3150 SEK
TCIAD0276-10G TCIAD0276-25G
1,2-Di(4-pyridyl)ethylene ≥98.0% (by GC, titration analysis)
1,2-Di(4-pyridyl)ethylene
Formel: C₁₂H₁₀N₂
MW: 182,23 g/mol
MDL Number: MFCD00006448
CAS nummer: 13362-78-2
EINECS: 236-432-3
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C