D (-)-Fruktsocker ≥99.0%

Leverantör: TCI EUROPE

Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose

F0060-500G F0060-25G
TCIAF0060-500GEA 375 SEK
TCIAF0060-500G TCIAF0060-25G
D (-)-Fruktsocker ≥99.0%
D (-)-Fruktsocker
Formel: C₆H₁₂O₆
MW: 180,16 g/mol
Kokpunkt: 552 °C (1013 hPa)
Smältpunkt: 100…110 °C
Densitet: 1,59 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00148910
CAS nummer: 57-48-7
EINECS: 200-333-3
Merck Index: 14,04273
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3708d5f7545f2c383536c255c43519688b7edc2c&vid=3708d5f7545f2c383536c255c43519688b7edc2c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (HPLC) (CAD) min. 99.0 area%
Specific rotation [a]20/D -90.0 to -94.0 deg (C=4, H2O) (after drying)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR