γ-Terpinene ≥95.0%
Leverantör: TCI EUROPE
Fara
Synonyms: gamma-Terpinene, p-Mentha-1,4-diene
M0318-25ML M0318-500ML
TCIAM0318-25MLEA 230 SEK
TCIAM0318-25ML TCIAM0318-500ML
γ-Terpinene ≥95.0%
γ-Terpinene
Formel: C₁₀H₁₆
MW: 136,24 g/mol
Kokpunkt: 183°C
Densitet: 0,846 g/cm³ (20 °C)
Flampunkt: 56°C
Storage Temperature: Kylskåp
MDL Number: MFCD00001537
CAS nummer: 99-85-4
EINECS: 202-794-6
UN: 2319
ADR: 3,III
Merck Index: 14,09170
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dea89c2e64f2214b9d6cf778b2ff9e9328b144d8&vid=dea89c2e64f2214b9d6cf778b2ff9e9328b144d8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %