trans-Decahydronaphthalene ≥98.0%
Leverantör: TCI
|
Fara
|
D0007-25ML
D0007-500ML
TCIAD0007-25MLEA
365
SEK
TCIAD0007-25ML
TCIAD0007-500ML
trans-Decahydronaphthalene ≥98.0%
trans-Decahydronaphthalene
Formel:
C₁₀H₁₈ Kokpunkt: 185°C Smältpunkt: –30 °C Densitet: 0,87 g/cm³ (20 °C) Flampunkt: 52°C Storage Temperature: Omgivande |
MDL Number:
MFCD00064191 CAS nummer: 493-02-7 EINECS: 207-771-4 UN: 1147 ADR: 3,III Merck Index: 14,02846 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...