4-Pyridinethiol ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Pyridine-4-thiol, 4-Mercaptopyridine
M0630-25G
M0630-5G
TCIAM0630-25GEA
2625
SEK
TCIAM0630-25G
TCIAM0630-5G
4-Pyridinethiol ≥97.0%
4-Pyridinethiol
Formel:
C₅H₅NS MW: 111,17 g/mol Smältpunkt: 186 °C Storage Temperature: Frys |
MDL Number:
MFCD00006422 CAS nummer: 4556-23-4 EINECS: 224-926-1 |
Specifikation testresultat
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...