Thioanisole ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene

T0191-25ML T0191-500ML T0191-100ML
TCIAT0191-25MLEA 215 SEK
TCIAT0191-25ML TCIAT0191-500ML TCIAT0191-100ML
Thioanisole ≥99.0%
Thioanisole
Formel: C₇H₈S
MW: 124,21 g/mol
Kokpunkt: 188°C
Smältpunkt: –15 °C
Densitet: 1,05 g/cm³ (20 °C)
Flampunkt: 57°C
Storage Temperature: Omgivande
MDL Number: MFCD00008559
CAS nummer: 100-68-5
EINECS: 202-878-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c362912d956b4063f3b387c96a6c59522aefe6e7&vid=c362912d956b4063f3b387c96a6c59522aefe6e7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR