1,2,4-Trihydroxibenzen ≥98.0%
Leverantör: TCI
|
Varning
|
H0249-10G
H0249-25G
TCIAH0249-10GEA
845
SEK
TCIAH0249-10G
TCIAH0249-25G
1,2,4-Trihydroxibenzen ≥98.0%
1,2,4-Trihydroxibenzen
Formel:
C₆H₆O₃ Kokpunkt: 362 °C (1013 hPa) Smältpunkt: 140 °C Densitet: 1,45 g/cm³ (20 °C) |
MDL Number:
MFCD00002198 CAS nummer: 533-73-3 EINECS: 208-575-1 Merck Index: 14,01073 |
|
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 145.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...