1,2,5-Pentanetriol ≥97.0%

Leverantör: TCI EUROPE
Varning

P1350-5G P1350-1G
TCIAP1350-5GEA 4000 SEK
TCIAP1350-5G TCIAP1350-1G
1,2,5-Pentanetriol ≥97.0%
1,2,5-Pentanetriol
Formel: C₅H₁₂O₃
MW: 120,15 g/mol
Kokpunkt: 195°C
MDL Number: MFCD01709258
CAS nummer: 14697-46-2
EINECS: 238-747-1

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR