2-metylresorcinol ≥98.0%

Leverantör: TCI
Fara

M0425-25G M0425-100G
TCIAM0425-25GEA 415 SEK
TCIAM0425-25G TCIAM0425-100G
2-metylresorcinol ≥98.0%
2-metylresorcinol
Formel: C₇H₈O₂
Kokpunkt: 264 °C (1013 hPa)
Smältpunkt: 120…122 °C
Flampunkt: 135 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002271
CAS nummer: 608-25-3
EINECS: 210-155-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fee3b7aee755c6f04ffaf47c9f29b82d958360f&vid=1fee3b7aee755c6f04ffaf47c9f29b82d958360f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR