(±)-1-Phenyl-1-propanol ≥98.0%
Leverantör: TCI
|
Varning
|
P0212-25G
P0212-500G
TCIAP0212-25GEA
310
SEK
TCIAP0212-25G
TCIAP0212-500G
(±)-1-Phenyl-1-propanol ≥98.0%
(±)-1-Phenyl-1-propanol
Formel:
C₆H₅CH(OH)CH₂CH₃ Kokpunkt: 212°C Densitet: 0,99 g/cm³ (20 °C) Flampunkt: 90°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004564 CAS nummer: 93-54-9 EINECS: 202-256-0 Merck Index: 14,03768 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...