Bis(2-etoxietyl)eter ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Ethoxyethyl ether, Diethyleneglycoldiethylether, Diethylene glycol diethyl ether

B0489-25ML B0489-500ML
TCIAB0489-25MLEA 215 SEK
TCIAB0489-25ML TCIAB0489-500ML
Bis(2-etoxietyl)eter ≥98.0%
Bis(2-etoxietyl)eter
Formel: C₈H₁₈O₃
MW: 162,23 g/mol
Kokpunkt: 188°C
Smältpunkt: –44°C
Densitet: 0,91 g/cm³ (20 °C)
Flampunkt: 71°C
Storage Temperature: Omgivande
MDL Number: MFCD00009254
CAS nummer: 112-36-7
EINECS: 203-963-7
Merck Index: 14,03118
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a134eeb8bc841df7ea77cdea019724a3d4cff84&vid=4a134eeb8bc841df7ea77cdea019724a3d4cff84&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR