3-Methoxythiophenol ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
3-Methoxybenzenethiol, 1-Methoxy-3-mercaptobenzene, 3-Mercaptoanisol, 3-Methoxybenzene-1-thiol, 3-Methoxyphenyl mercaptan, m-Methoxythiophenol
M1194-25G
M1194-5G
TCIAM1194-25GEA
1675
SEK
TCIAM1194-25G
TCIAM1194-5G
3-Methoxythiophenol ≥98.0%
3-Methoxythiophenol
Formel:
C₇H₈OS Kokpunkt: 114°C Densitet: 1,136 g/cm³ (20 °C) Flampunkt: 96°C |
MDL Number:
MFCD00004841 CAS nummer: 15570-12-4 EINECS: 239-617-7 UN: 2810 ADR: 6.1,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...