Eugenol ≥99.0%

Leverantör: TCI
Fara

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

A0232-25ML A0232-500ML A0232-100ML
TCIAA0232-25MLEA 215 SEK
TCIAA0232-25ML TCIAA0232-500ML TCIAA0232-100ML
Eugenol ≥99.0%
Eugenol
Formel: C₁₀H₁₂O₂
MW: 164.2 g/mol
Kokpunkt: 255
Smältpunkt: -10
Flampunkt: 127°C
Storage Temperature: Omgivande
MDL Number: MFCD00008654
CAS nummer: 97-53-0
EINECS: 202-589-1
Merck Index: 14,03898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=957e7fb834727da996ed88d493a82a3cd91b8c95&vid=957e7fb834727da996ed88d493a82a3cd91b8c95&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR