1-Ethoxynaphthalene ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Ethyl 1-naphthyl ether, 1-Ehoxynaphthalene

E0532-25G E0532-5G
TCIAE0532-25GEA 1700 SEK
TCIAE0532-25G TCIAE0532-5G
1-Ethoxynaphthalene ≥95.0%
1-Ethoxynaphthalene
Formel: C₁₂H₁₂O
MW: 172,23 g/mol
Kokpunkt: 280 °C (1013 hPa)
Smältpunkt: 5,5 °C
Densitet: 1,0711 g/cm³ (4 °C)
MDL Number: MFCD00003928
CAS nummer: 5328-01-8
EINECS: 226-213-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dc160387f2ac729cde110bcfa193cf80aeccfec&vid=5dc160387f2ac729cde110bcfa193cf80aeccfec&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Brown clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 1.0570 to 1.0670

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR