Trimethyl orthopropionate ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
T2288-25G
T2288-5G
TCIAT2288-25GEA
1150
SEK
TCIAT2288-25G
TCIAT2288-5G
Trimethyl orthopropionate ≥97.0%
Trimethyl orthopropionate
Formel:
C₆H₁₄O₃ MW: 134,18 g/mol Kokpunkt: 128°C Flampunkt: 19°C |
MDL Number:
MFCD00043305 CAS nummer: 24823-81-2 UN: 3272 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...