2-Methyl-5-nitroanisole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Methoxy-2-methyl-5-nitrobenzene, Methyl 5-nitro-o-tolyl ether, 2-Methoxy-4-nitrotoluene
M1681-25G
M1681-500G
TCIAM1681-25GEA
415
SEK
TCIAM1681-25G
TCIAM1681-500G
2-Methyl-5-nitroanisole ≥98.0%
2-Methyl-5-nitroanisole
Formel:
C₈H₉NO₃ MW: 167,16 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00043912 CAS nummer: 13120-77-9 EINECS: 236-048-6 ADR: 6.1,III |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 70.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...