4-Nitroimidazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole
N0506-25G
N0506-500G
TCIAN0506-25GEA
310
SEK
TCIAN0506-25G
TCIAN0506-500G
4-Nitroimidazole ≥98.0%
4-Nitroimidazole
Formel:
C₃H₃N₃O₂ MW: 113,08 g/mol Smältpunkt: 312°C Storage Temperature: Omgivande |
MDL Number:
MFCD00005196 CAS nummer: 3034-38-6 EINECS: 221-224-7 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...