Diisobutylamine
Leverantör: TCI EUROPE
|
Fara
|
D0614-25ML
D0614-500ML
TCIAD0614-25MLEA
215
SEK
TCIAD0614-25ML
TCIAD0614-500ML
Diisobutylamine
Diisobutylamine
Formel:
C₈H₁₉N MW: 129,25 g/mol Kokpunkt: 139°C Smältpunkt: –74°C Densitet: 0,763 g/cm³ (0 °C) Flampunkt: 29°C |
MDL Number:
MFCD00008930 CAS nummer: 110-96-3 EINECS: 203-819-3 UN: 2361 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...