m-Fenylendiamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1,3-Fenylendiamin
P1911-25G
P1911-250G
TCIAP1911-25GEA
535
SEK
TCIAP1911-25G
TCIAP1911-250G
m-Fenylendiamin ≥98.0% (by GC, titration analysis)
m-Fenylendiamin
Formel:
C₆H₈N₂ MW: 108,14 g/mol Kokpunkt: 287°C Smältpunkt: 63…64 °C Densitet: 1,14 g/cm³ (20 °C) Flampunkt: 187 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00007799 CAS nummer: 108-45-2 EINECS: 203-584-7 UN: 1673 ADR: 6.1,III Merck Index: 14,07283 |
Specifikation testresultat
Appearance | White to Almost white flake |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...