m-Fenylendiamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: 1,3-Fenylendiamin
P1892-25G P1892-500G
TCIAP1892-25GEA 245 SEK
TCIAP1892-25G TCIAP1892-500G
m-Fenylendiamin ≥98.0% (by GC, titration analysis)
m-Fenylendiamin
Formel: C₆H₈N₂
MW: 108.14 g/mol
Kokpunkt: 287°C
Flampunkt: 187
Storage Temperature: Omgivande
MDL Number: MFCD00007799
CAS nummer: 108-45-2
EINECS: 203-584-7
UN: 1673
ADR: 6.1 III
Merck Index: 14,07283
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d0216b0b14bc9bfa27be7a06f46d1036eba7662&vid=6d0216b0b14bc9bfa27be7a06f46d1036eba7662&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 62.0 to 66.0 deg-C