1,8-Naftalendiamin ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1,8-naphthylenediamine, 1,8-Naphthalenediamine, Naphthalene-1,8-diamine
D0102-25G D0102-500G D0102-100G
TCIAD0102-25GEA 405 SEK
TCIAD0102-25G TCIAD0102-500G TCIAD0102-100G
1,8-Naftalendiamin ≥98.0%
1,8-Naftalendiamin
Formel: C₁₀H₁₀N₂
MW: 158,2 g/mol
Kokpunkt: 352 °C (1013 hPa)
Smältpunkt: 64…65 °C
Densitet: 1,18 g/cm³ (20 °C)
Flampunkt: 195 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004033
CAS nummer: 479-27-6
EINECS: 207-529-8
ADR: 8,III
Merck Index: 14,06372
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22a7dd73ca0dfb767c8f65b86ef444ff7d78c29d&vid=22a7dd73ca0dfb767c8f65b86ef444ff7d78c29d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Red to Dark blue to Black powder to lump
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 63.0 to 67.0 deg-C