4-Nitro-o-toluidin ≥98.0%
Leverantör: TCI
Fara
Synonyms: 4-Nitro-2-methylaniline, 2-Methyl-4-nitroaniline, 2-Amino-5-nitrotoluene
N0116-25G N0116-500G
TCIAN0116-25GEA 815 SEK
TCIAN0116-25G TCIAN0116-500G
4-Nitro-o-toluidin ≥98.0%
4-Nitro-o-toluidin
Formel: C₇H₈N₂O₂
MW: 152.15 g/mol
Smältpunkt: 135
Storage Temperature: Omgivande
MDL Number: MFCD00007734
CAS nummer: 99-52-5
EINECS: 202-762-1
UN: 2660
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd1f267f9e2887ea02cd10db6a5fd02c58b85bd6&vid=cd1f267f9e2887ea02cd10db6a5fd02c58b85bd6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 133.0 deg-C
Solubility in hot Methanol almost transparency