Adenin ≥99.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
A0149-25G A0149-250G
TCIAA0149-25GEA 270 SEK
TCIAA0149-25G TCIAA0149-250G
Adenin ≥99.0% (by HPLC, titration analysis)
Adenin
Formel: C₅H₅N₅
MW: 135,13 g/mol
Smältpunkt: 365°C
Densitet: 0,99741 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00041790
CAS nummer: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,00152
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc9c05d0556f054aba80d7b122333c5f4f9eeeb&vid=cbc9c05d0556f054aba80d7b122333c5f4f9eeeb&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in hot 1mol/L HCl almost transparency