m-Toluidin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: 3-Toluidin
T0298-25ML T0298-500ML
TCIAT0298-25MLEA 245 SEK
TCIAT0298-25ML TCIAT0298-500ML
m-Toluidin ≥98.0% (by GC, titration analysis)
m-Toluidin
Formel: C₇H₉N
MW: 107.16 g/mol
Kokpunkt: 203°C
Smältpunkt: -30
Flampunkt: 86°C
Storage Temperature: Kylskåp
MDL Number: MFCD00007808
CAS nummer: 108-44-1
EINECS: 203-583-1
UN: 1708
ADR: 6.1 II
Merck Index: 14,09536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e753bf6f27b453867b1daa903484c4e47b96f8&vid=c6e753bf6f27b453867b1daa903484c4e47b96f8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %