5-Fluoro-2-nitroanisole ≥98.0%
Leverantör: TCI
Varning
Synonyms: 4-Fluoro-2-methoxy-1-nitrobenzene
F0703-25G F0703-5G
TCIAF0703-25GEA 1700 SEK
TCIAF0703-25G TCIAF0703-5G
5-Fluoro-2-nitroanisole ≥98.0%
5-Fluoro-2-nitroanisole
Formel: C₇H₆FNO₃
Kokpunkt: 272°C
Storage Temperature: Omgivande
MDL Number: MFCD00077541
CAS nummer: 448-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a931226b6748fb7ac87d305a2b64433f69e01b7&vid=0a931226b6748fb7ac87d305a2b64433f69e01b7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 52.0 deg-C