Trifluoroacetamide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,2,2-Trifluoroacetamide
T0598-25G
T0598-500G
T0598-100G
TCIAT0598-25GEA
285
SEK
TCIAT0598-25G
TCIAT0598-500G
TCIAT0598-100G
Trifluoroacetamide ≥98.0%
Trifluoroacetamide
Formel:
C₂H₂F₃NO MW: 113,04 g/mol Kokpunkt: 163°C Smältpunkt: 73…75 °C Densitet: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00008008 CAS nummer: 354-38-1 EINECS: 206-559-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 96.0 % |
Melting point | 72.0 to 75.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...