3'-(Trifluoromethyl)acetophenone ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
T1546-25G T1546-5G
TCIAT1546-25GEA 1300 SEK
TCIAT1546-25G TCIAT1546-5G
3'-(Trifluoromethyl)acetophenone ≥97.0%
3'-(Trifluoromethyl)acetophenone
Formel: C₉H₇F₃O
MW: 188,15 g/mol
Kokpunkt: 199°C
Densitet: 1,23 g/cm³ (20 °C)
Flampunkt: 83°C
Storage Temperature: Omgivande
MDL Number: MFCD00000391
CAS nummer: 349-76-8
EINECS: 206-490-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b72df896ae0192a0316d5ff547fa23506023255&vid=7b72df896ae0192a0316d5ff547fa23506023255&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %