α,α,α-Trifluoro-3-tolualdehyde ≥95.0%

Leverantör: TCI
Varning

Synonyms: 3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23

T1399-25G T1399-5G
TCIAT1399-25GEA 1025 SEK
TCIAT1399-25G TCIAT1399-5G
α,α,α-Trifluoro-3-tolualdehyde ≥95.0%
α,α,α-Trifluoro-3-tolualdehyde
Formel: C₈H₅F₃O
Kokpunkt: 66°C
Densitet: 1,3024 g/cm³ (20 °C)
Flampunkt: 68°C
MDL Number: MFCD00003373
CAS nummer: 454-89-7
EINECS: 207-228-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4837108d8a6741f576cc0acac8045961cd0f5aae&vid=4837108d8a6741f576cc0acac8045961cd0f5aae&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR