(3-Nitrophenyl)acetic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

N0442-1G N0442-5G
TCIAN0442-1GEA 430 SEK
TCIAN0442-1G TCIAN0442-5G
(3-Nitrophenyl)acetic acid ≥98.0%
(3-Nitrophenyl)acetic acid
Formel: (C₆H₄(NO₂))CH₂COOH
MW: 181,15 g/mol
Smältpunkt: 120…122 °C
MDL Number: MFCD00007278
CAS nummer: 1877-73-2
EINECS: 217-512-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84692e9b81f1f15395321e1514b32dab856e2da6&vid=84692e9b81f1f15395321e1514b32dab856e2da6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light red to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 121.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR