Barium stearate

Leverantör: TCI
Varning

Synonyms: Stearic acid barium salt

S0235-25G S0235-500G
TCIAS0235-25GEA 215 SEK
TCIAS0235-25G TCIAS0235-500G
Barium stearate
Barium stearate
Formel: C₃₆H₇₀BaO₄
MW: 704.28 g/mol
Smältpunkt: 120°C
Storage Temperature: Omgivande
MDL Number: MFCD00050711
CAS nummer: 6865-35-6
EINECS: 229-966-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=867ee4ef63897ad96054e83cbfc28d3af0cbfe6b&vid=867ee4ef63897ad96054e83cbfc28d3af0cbfe6b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Stearic acid (composition of fatty acid) min. 60.0 %
Content (Barium) 18.5 to 20.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR