Nikotinsyra (Niacin) ≥99.0% (by HPLC, titration analysis)
Leverantör: TCI
Varning
Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3
N0082-25G N0082-500G
TCIAN0082-25GEA 215 SEK
TCIAN0082-25G TCIAN0082-500G
Nikotinsyra (Niacin) ≥99.0% (by HPLC, titration analysis)
Nikotinsyra (Niacin)
Formel: C₆H₅NO₂
Kokpunkt: 258 °C (1013 hPa)
Smältpunkt: 142 °C
Densitet: 1,473 g/cm³ (20 °C)
Flampunkt: 193 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006391
CAS nummer: 59-67-6
EINECS: 200-441-0
Merck Index: 14,06525
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dceda564a5233e15ebe17c9453abc181376a5fd7&vid=dceda564a5233e15ebe17c9453abc181376a5fd7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 235.0 to 240.0 deg-C
Solubility in hot Water almost transparency