(±)-2-Methylvaleric acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: (±)-2-Methylpentanoic acid, 2-Methylpentanoic Acid, 2-Methylvaleric Acid
M0610-25ML M0610-500ML
TCIAM0610-25MLEA 215 SEK
TCIAM0610-25ML TCIAM0610-500ML
(±)-2-Methylvaleric acid ≥98.0% (by GC, titration analysis)
(±)-2-Methylvaleric acid
Formel: C₆H₁₂O₂
MW: 116.16 g/mol
Kokpunkt: 195°C
Smältpunkt: -85
Flampunkt: 107°C
MDL Number: MFCD00002671
CAS nummer: 97-61-0
EINECS: 202-594-9
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f35a528d6bd5e8d5b3acf34b5afcb7adb2edc7cd&vid=f35a528d6bd5e8d5b3acf34b5afcb7adb2edc7cd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %