Dietylmaleat ≥90.0%

Leverantör: TCI
Varning

Synonyms: Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate

M0010-25ML M0010-500ML
TCIAM0010-25MLEA 215 SEK
TCIAM0010-25ML TCIAM0010-500ML
Dietylmaleat ≥90.0%
Dietylmaleat
Formel: C₈H₁₂O₄
Kokpunkt: 225 °C (1013 hPa)
Smältpunkt: –11 °C
Densitet: 1,07 g/cm³ (20 °C)
Flampunkt: 107°C
Storage Temperature: Omgivande
MDL Number: MFCD00009191
CAS nummer: 141-05-9
EINECS: 205-451-9
Merck Index: 14,03123
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a924600a8f4016093dd37e86f61e14f908306196&vid=a924600a8f4016093dd37e86f61e14f908306196&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR