Dimetylkarbonat ≥98.0%

Leverantör: TCI
Fara

C0053-500ML C0053-25ML C0053-100ML
TCIAC0053-500MLEA 645 SEK
TCIAC0053-500ML TCIAC0053-25ML TCIAC0053-100ML
Dimetylkarbonat ≥98.0%
Dimetylkarbonat
Formel: C₃H₆O₃
Kokpunkt: 90°C
Smältpunkt: 4°C
Densitet: 1,07 g/cm³ (20 °C)
Flampunkt: 17°C
Storage Temperature: Omgivande
MDL Number: MFCD00008420
CAS nummer: 616-38-6
EINECS: 210-478-4
UN: 1161
ADR: 3,II
Merck Index: 14,03241
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b80c437ea2c9a7fb10d6cee82a8793c0c0e998a7&vid=b80c437ea2c9a7fb10d6cee82a8793c0c0e998a7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR