3-Nitrobenzoyl chloride ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: m-Nitrobenzoyl chloride
N0174-25G N0174-500G
TCIAN0174-25GEA 285 SEK
TCIAN0174-25G TCIAN0174-500G
3-Nitrobenzoyl chloride ≥98.0% (by GC, titration analysis)
3-Nitrobenzoyl chloride
Formel: C₇H₄ClNO₃
MW: 185,57 g/mol
Kokpunkt: 275 °C (1013 hPa)
Smältpunkt: 30…33 °C
Densitet: 1,42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007247
CAS nummer: 121-90-4
EINECS: 204-505-9
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de501305e3b3547ca656e3a8b344384f42aafd4b&vid=de501305e3b3547ca656e3a8b344384f42aafd4b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Toluene very faint turbidity