4,4'-Dimethoxybenzil ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione
A1028-25G A1028-5G
TCIAA1028-25GEA 1400 SEK
TCIAA1028-25G TCIAA1028-5G
4,4'-Dimethoxybenzil ≥98.0%
4,4'-Dimethoxybenzil
Formel: C₁₆H₁₄O₄
MW: 270,28 g/mol
Kokpunkt: 215 - 219 °C (3 -rr)
Smältpunkt: 128…131 °C
Densitet: 1,36 g/cm³ (20 °C)
MDL Number: MFCD00008405
CAS nummer: 1226-42-2
EINECS: 214-960-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22d377db000010e2f7650bb0089f95d43774eb6b&vid=22d377db000010e2f7650bb0089f95d43774eb6b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 132.0 to 135.0 deg-C