7-Hydroxycoumarin 98,0 GC_ANALYSMETOD
Leverantör: TCI
Varning
Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
H0236-25G H0236-5G
TCIAH0236-25GEA 630 SEK
TCIAH0236-25G TCIAH0236-5G
7-Hydroxycoumarin 98,0 GC_ANALYSMETOD
7-Hydroxycoumarin
Formel: C₉H₆O₃
MW: 162.14 g/mol
Kokpunkt: 382
Storage Temperature: Omgivande
MDL Number: MFCD00006878
CAS nummer: 93-35-6
EINECS: 202-240-3
Merck Index: 14,09847
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %