4-Nitrobenzophenone ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: p-Nitrobenzophenone
N0365-10G N0365-25G
TCIAN0365-10GEA 335 SEK
TCIAN0365-10G TCIAN0365-25G
4-Nitrobenzophenone ≥99.0%
4-Nitrobenzophenone
Formel: C₁₃H₉NO₃
MW: 227,22 g/mol
Smältpunkt: 136…137 °C
Densitet: 1,406 g/cm³ (0 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007354
CAS nummer: 1144-74-7
EINECS: 214-542-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e48ed9ec52c23ac28e7fa51781bdac57f10f252&vid=2e48ed9ec52c23ac28e7fa51781bdac57f10f252&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 137.0 to 140.0 deg-C
Solubility in hot EtOH almost transparency