3-Hexyne-2-one ≥95.0%
Leverantör: TCI
|
Fara
|
H0741-1ML
H0741-5ML
TCIAH0741-1MLEA
550
SEK
TCIAH0741-1ML
TCIAH0741-5ML
3-Hexyne-2-one ≥95.0%
3-Hexyne-2-one
Formel:
C₆H₈O Kokpunkt: 77°C Densitet: 0,88 g/cm³ (20 °C) Flampunkt: 39°C |
MDL Number:
MFCD00041627 CAS nummer: 1679-36-3 UN: 1992 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...