Propiononitrile ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Propionitrile

P0514-25ML P0514-500ML P0514-100ML
TCIAP0514-25MLEA 230 SEK
TCIAP0514-25ML TCIAP0514-500ML TCIAP0514-100ML
Propiononitrile ≥99.0%
Propiononitrile
Formel: C₃H₅N
MW: 55,08 g/mol
Kokpunkt: 96°C
Smältpunkt: –93 °C
Densitet: 0,782 g/cm³ (20 °C)
Flampunkt: 13°C
Storage Temperature: Omgivande
MDL Number: MFCD00001948
CAS nummer: 107-12-0
EINECS: 203-464-4
UN: 2404
ADR: 3,II
Merck Index: 14,07827
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b00b09f22385bb28657bf06c2250c61397c162e2&vid=b00b09f22385bb28657bf06c2250c61397c162e2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR