3-Chloro-2-methoxypyridine ≥97.0%
Leverantör: TCI
|
Varning
|
C2565-1G
C2565-5G
TCIAC2565-1GEA
375
SEK
TCIAC2565-1G
TCIAC2565-5G
3-Chloro-2-methoxypyridine ≥97.0%
3-Chloro-2-methoxypyridine
Formel:
C₆H₆ClNO MW: 143.57 g/mol Kokpunkt: 53°C |
MDL Number:
MFCD03095227 CAS nummer: 13472-84-9 UN: 1993 ADR: 3 III |
|
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...